4-(Methoxycarbonyl)cyclohexanecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1,4-Cyclohexanedicarboxylic acid monomethyl ester

A113751-100G A113751-25G A113751-5G
77284-316EA 240 USD
77284-316 77284-318 77284-320
4-(Methoxycarbonyl)cyclohexanecarboxylic acid 95%
4-(Methoxycarbonyl)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄O₄
MDL Number: MFCD06797561
CAS Number: 32529-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR