2-Hydroxyethyl methyl sulfone 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylsulphonyl) ethanol, 2-Methylsulphonylethanol

A113743-100G A113743-1G A113743-25G A113743-5G
77245-832EA 369.77 USD
77245-832 77245-834 77245-836 77245-838
2-Hydroxyethyl methyl sulfone 97%
2-Hydroxyethyl methyl sulfone

Precaución: for Research Use Only

Formula: C₃H₈O₃S
MW: 124.15 g/mol
MDL Number: MFCD00014747
CAS Number: 15205-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR