9-(4-Bromophenyl)-9-phenylfluorene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
9-(4-Bromophenyl)-9-phenyl-9H-fluorene
A113723-100G
A113723-1G
A113723-250MG
A113723-25G
A113723-500G
A113723-5G
77233-760EA
442.9
USD
77233-760
77233-762
77233-764
77233-766
77233-768
77233-770
9-(4-Bromophenyl)-9-phenylfluorene 97%
9-(4-Bromophenyl)-9-phenylfluorene
Precaución: for Research Use Only
Formula:
C₂₅H₁₇Br MW: 397.32 g/mol |
MDL Number:
MFCD28975087 CAS Number: 937082-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...