(R)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride 97%

Supplier: Ambeed
Warning

A113684-1G A113684-5G
77245-828EA 207.03 USD
77245-828 77245-830
(R)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride 97%
(R)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₀ClNO₄S
MW: 369.86 g/mol
MDL Number: MFCD30570304
CAS Number: 2049127-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR