Napabucasin 98%

Supplier: Ambeed
Warning

Synonyms: Napabucasin, 2-Acetylnaphtho[2,3-b]furan-4,9-dione

A113651-10MG A113651-1MG A113651-250MG A113651-5MG A113651-100MG A113651-1G
77233-750EA 37.08 USD
77233-750 77233-752 77233-754 77233-756 77614-888 77614-890
Napabucasin 98%
Napabucasin

Precaución: for Research Use Only

Formula: C₁₄H₈O₄
MW: 240.21 g/mol
MDL Number: MFCD28155270
CAS Number: 83280-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR