5-(2,3-Dichlorophenyl)-1,3,4-thiadiazol-2-amine 97%

Supplier: Ambeed
Warning

A113585-10G A113585-1G A113585-250MG A113585-25G A113585-5G
77233-730EA 317.01 USD
77233-730 77233-732 77233-734 77233-736 77233-738
5-(2,3-Dichlorophenyl)-1,3,4-thiadiazol-2-amine 97%
5-(2,3-Dichlorophenyl)-1,3,4-thiadiazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N₃S
MDL Number: MFCD02664063
CAS Number: 299443-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR