4-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A113575-100MG A113575-1G A113575-250MG A113575-25G A113575-5G
77284-296EA 6 USD
77284-296 77284-298 77284-300 77284-302 77284-304
4-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one 95%
4-Bromo-6-fluoro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆BrFO
MDL Number: MFCD09038174
CAS Number: 174603-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR