2-(4-Benzoyl-3-hydroxyphenoxy)ethyl acrylate 97%

Supplier: Ambeed
Warning

A113525-100G A113525-1G A113525-25G A113525-5G
77233-716EA 146.26 USD
77233-716 77233-718 77233-720 77233-722
2-(4-Benzoyl-3-hydroxyphenoxy)ethyl acrylate 97%
2-(4-Benzoyl-3-hydroxyphenoxy)ethyl acrylate

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₅
MW: 312.31 g/mol
MDL Number: MFCD00081133
CAS Number: 16432-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR