2-Amino-5-bromo-4-methyl-3-nitropyridine 95%

Supplier: Ambeed
Warning

A113510-10G A113510-1G A113510-250MG A113510-5G A113510-25G
77245-816EA 20 USD
77245-816 77245-818 77245-820 77245-822 77745-841
2-Amino-5-bromo-4-methyl-3-nitropyridine 95%
2-Amino-5-bromo-4-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O₂
MDL Number: MFCD00092197
CAS Number: 100367-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR