5-Nitro-1H-indazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Nitro-1H-indazole, 5-NINA, 5-Nitroindazole

A113492-100G A113492-10G A113492-25G A113492-500G A113492-5G
77271-048EA 42 USD
77271-048 77271-050 77271-052 77271-054 77271-056
5-Nitro-1H-indazole 98%
5-Nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.14 g/mol
MDL Number: MFCD00005693
CAS Number: 5401-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR