4'-Chloro-2'-fluoroacetanilide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Chloro-2-fluorophenyl)acetamide

A113382-100G A113382-25G A113382-5G
77284-284EA 154.5 USD
77284-284 77284-286 77284-288
4'-Chloro-2'-fluoroacetanilide 98%
4'-Chloro-2'-fluoroacetanilide

Precaución: for Research Use Only

Formula: C₈H₇ClFNO
MW: 187.6 g/mol
MDL Number: MFCD00042520
CAS Number: 59280-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR