2-Phenylpyridin-3-ol 95%

Supplier: Ambeed
Warning

A113349-100MG A113349-1G A113349-250MG A113349-5G
77271-030EA 22 USD
77271-030 77271-032 77271-034 77271-036
2-Phenylpyridin-3-ol 95%
2-Phenylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MDL Number: MFCD00731184
CAS Number: 3308-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR