2-Bromo-4-methylnicotinonitrile 95%

Supplier: Ambeed
Warning

A113271-100MG A113271-250MG
77257-786EA 20.6 USD
77257-786 77257-788
2-Bromo-4-methylnicotinonitrile 95%
2-Bromo-4-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.032 g/mol
MDL Number: MFCD09834634
CAS Number: 65996-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR