Didodecyl hydrogen phosphate 97%

Supplier: Ambeed
Warning

A113247-100MG A113247-1G A113247-250MG
77257-780EA 41.2 USD
77257-780 77257-782 77257-784
Didodecyl hydrogen phosphate 97%
Didodecyl hydrogen phosphate

Precaución: for Research Use Only

Formula: C₂₄H₅₁O₄P
MW: 434.63 g/mol
MDL Number: MFCD00015070
CAS Number: 7057-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR