3-Bromo-5-ethoxy-4-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A113182-10G A113182-1G
77257-774EA 66 USD
77257-774 77257-776
3-Bromo-5-ethoxy-4-hydroxybenzaldehyde 97%
3-Bromo-5-ethoxy-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD00173904
CAS Number: 3111-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR