2,4-Dichloro-5-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A113164-250MG A113164-25G A113164-5G
77245-804EA 7 USD
77245-804 77245-806 77245-808
2,4-Dichloro-5-fluorobenzaldehyde 98%
2,4-Dichloro-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
MDL Number: MFCD01631379
CAS Number: 86522-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR