(2-Bromo-5-methoxyphenyl)methanol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-methoxybenzyl alcohol
A113087-1G
A113087-25G
A113087-5G
77257-768EA
43.26
USD
77257-768
77257-770
77257-772
(2-Bromo-5-methoxyphenyl)methanol 95%
(2-Bromo-5-methoxyphenyl)methanol
Precaución: for Research Use Only
Formula:
C₈H₉BrO₂ MW: 217.05 g/mol |
MDL Number:
MFCD03695463 CAS Number: 150192-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...