3-(Trimethylsilyl)propiolic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Trimethylsilyl)propynoic acid

A113060-100MG A113060-1G A113060-250MG A113060-25G A113060-5G
77271-004EA 12 USD
77271-004 77271-006 77271-008 77271-010 77271-012
3-(Trimethylsilyl)propiolic acid 95%
3-(Trimethylsilyl)propiolic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂Si
MDL Number: MFCD00190216
CAS Number: 5683-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR