(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S,R,S)-AHPC monohydrochloride
A113059-1MG
A113059-25MG
A113059-50MG
A113059-5G
A113059-5MG
A113059-1G
A113059-250MG
77257-758EA
5.15
USD
77257-758
77257-760
77257-762
77257-764
77257-766
77549-196
77565-984
(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride 98%
(2S,4R)-1-((S)-2-Amino-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride
Precaución: For research use only.
Formula:
C₂₂H₃₁ClN₄O₃S MW: 467.02 g/mol |
MDL Number:
MFCD30502992 CAS Number: 1448189-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...