Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate 99%

Supplier: Ambeed
Warning

Synonyms: Tris(2,2'-bipyridine)ruthenium bis(hexafluorophosphate), Ruthenium-tris(2,2′-bipyridyl) dihexafluorophosphate, Ru(bpy)₃(PF₆)₂, Tris(2,2'-bipyridine)ruthenium(II) Bis(hexafluorophosphate)

A113049-100MG A113049-1G A113049-250MG A113049-5G
77245-798EA 20 USD
77245-798 77245-800 77245-802 77614-882
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate 99%
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₀H₂₄F₁₂N₆P₂Ru
MW: 859.562 g/mol
MDL Number: MFCD11042502
CAS Number: 60804-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR