(2R,3R,4S,5R)-2-(2-Chloro-6-methoxy-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 96%

Supplier: Ambeed
Warning

A113028-100MG A113028-1G A113028-250MG
77233-692EA 18.54 USD
77233-692 77233-694 77233-696
(2R,3R,4S,5R)-2-(2-Chloro-6-methoxy-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 96%
(2R,3R,4S,5R)-2-(2-Chloro-6-methoxy-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₄O₅
MW: 316.70 g/mol
MDL Number: MFCD28579842
CAS Number: 15465-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR