4-Fluoro-5-nitrobenzene-1,2-diamine 97%
Supplier: Ambeed
|
Warning
|
A112850-1G
A112850-250MG
A112850-5G
77284-262EA
55
USD
77284-262
77284-264
77284-266
4-Fluoro-5-nitrobenzene-1,2-diamine 97%
4-Fluoro-5-nitrobenzene-1,2-diamine
Precaución: for Research Use Only
Formula:
C₆H₆FN₃O₂ |
MDL Number:
MFCD00828924 CAS Number: 113269-06-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...