(S)-tert-Butyl (1-(3-bromophenyl)-2-hydroxyethyl)carbamate 97%

Supplier: Ambeed
Warning

A112692-100MG A112692-1G A112692-5G A112692-250MG
77233-664EA 20 USD
77233-664 77233-666 77233-668 77745-835
(S)-tert-Butyl (1-(3-bromophenyl)-2-hydroxyethyl)carbamate 97%
(S)-tert-Butyl (1-(3-bromophenyl)-2-hydroxyethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₃
MDL Number: MFCD27952001
CAS Number: 910308-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR