Altretamine 97%

Supplier: Ambeed
Warning

Synonyms: Hexamethylmelamine, NSC -13875, 2,4,6-Tris(dimethylamino)-1,3,5-triazine Hexamethylmelamine, N2,N2,N4,N4,N6,N6-hexamethyl-1,3,5-triazine-2,4,6-triamine

A112657-100MG A112657-1G A112657-250MG A112657-25MG A112657-1MG A112657-5G
77245-782EA 10.3 USD
77245-782 77245-784 77245-786 77245-788 77745-833 77745-834
Altretamine 97%
Altretamine

Precaución: for Research Use Only

Formula: C₉H₁₈N₆
MW: 210.28 g/mol
MDL Number: MFCD00549245
CAS Number: 645-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR