N-Phenylpiperidin-4-one 97%

Supplier: Ambeed
Warning

A112650-100MG A112650-1G A112650-250MG A112650-5G A112650-10G A112650-25G
77233-648EA 14.42 USD
77233-648 77233-650 77233-652 77233-654 77745-831 77745-832
N-Phenylpiperidin-4-one 97%
N-Phenylpiperidin-4-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MW: 175.22 g/mol
MDL Number: MFCD07367770
CAS Number: 19125-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR