6-(4-Aminostyryl)picolinic acid 98%

Supplier: Ambeed
Warning

A112587-10MG A112587-1MG A112587-25MG A112587-50MG A112587-5MG
77270-978EA 117 USD
77270-978 77270-980 77270-982 77270-984 77270-986
6-(4-Aminostyryl)picolinic acid 98%
6-(4-Aminostyryl)picolinic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₂
MDL Number: MFCD05999206
CAS Number: 115453-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR