4-Methoxyphenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)anisole, 2-(4-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methoxyphenylboronic acid Pinacol ester

A112414-100G A112414-10G A112414-1G A112414-25G A112414-5G
77284-230EA 147 USD
77284-230 77284-232 77284-234 77284-236 77284-238
4-Methoxyphenylboronic acid pinacol ester 97%
4-Methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃
MW: 234.1 g/mol
MDL Number: MFCD05663864
CAS Number: 171364-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR