U-93631 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4,4-dimethyl-4,5-dihydroimidazo[1,5-a]quinoxaline-3-carboxylate, 4,5-Dihydro-4,4-dimethyl-imidazo[1,5-a]quinoxaline-3-carboxylic acid 1,1-dimethylethyl ester, U93631

A112278-100MG A112278-10MG A112278-1MG A112278-50MG A112278-5MG
77257-714EA 267 USD
77257-714 77257-716 77257-718 77257-720 77257-722
U-93631 98%
U-93631

Precaución: for Research Use Only

Formula: C₁₇H₂₁N₃O₂
MW: 299.37 g/mol
MDL Number: MFCD10566712
CAS Number: 152273-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR