Lumiracoxib 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-((2-chloro-6-fluorophenyl)amino)-5-methylphenyl)acetic acid, CGS-35189, COX-189,  

A112255-100MG A112255-10MG A112255-1MG A112255-50MG
77245-750EA 118 USD
77245-750 77245-752 77245-754 77245-756
Lumiracoxib 98%
Lumiracoxib

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClFNO₂
MDL Number: MFCD07186254
CAS Number: 220991-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR