(2,2'-Bipyridine)diiodonickel 98%

Supplier: Ambeed
Danger

A112195-1G A112195-5G
77257-710EA 105 USD
77257-710 77257-712
(2,2'-Bipyridine)diiodonickel 98%
(2,2'-Bipyridine)diiodonickel

Precaución: for Research Use Only

Formula: C₁₀H₈I₂N₂Ni
MDL Number: MFCD32672292
CAS Number: 59448-25-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR