2-Coumaranone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one
A112172-1000G
A112172-100G
A112172-10G
A112172-25G
A112172-500G
77233-626EA
106
USD
77233-626
77233-628
77233-630
77233-632
77614-870
2-Coumaranone 98%
2-Coumaranone
Precaución: for Research Use Only
Formula:
C₈H₆O₂ |
MDL Number:
MFCD00005856 CAS Number: 553-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...