2-((Difluoromethyl)thio)benzo[d]thiazole 98%

Supplier: Ambeed
Warning

A111881-1G A111881-250MG A111881-5G
77233-616EA 146.26 USD
77233-616 77233-618 77233-620
2-((Difluoromethyl)thio)benzo[d]thiazole 98%
2-((Difluoromethyl)thio)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₅F₂NS₂
MW: 217.25 g/mol
MDL Number: MFCD00518584
CAS Number: 943-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR