4-Phenylpyridin-2-ol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxy-4-phenylpyridine
A111730-1G
A111730-250MG
77257-692EA
189.52
USD
77257-692
77257-694
4-Phenylpyridin-2-ol 95%
4-Phenylpyridin-2-ol
Precaución: for Research Use Only
Formula:
C₁₁H₉NO MW: 171.1953 g/mol |
MDL Number:
MFCD07437954 CAS Number: 19006-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...