3-Oxoindane-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A111678-100MG A111678-1G A111678-250MG
77233-602EA 36 USD
77233-602 77233-604 77233-606
3-Oxoindane-5-carboxylic acid 97%
3-Oxoindane-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
MDL Number: MFCD02179295
CAS Number: 60031-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR