3,4',5-Trimethoxy-trans-stilbene 98%

Supplier: Ambeed
Danger

Synonyms: trans-1-(3,5-Dimethoxyphenyl)-2-(4-methoxyphenyl)ethylene, (E)-1,3-Dimethoxy-5-(4-methoxystyryl)benzene, trans-Trimethoxyresveratrol

A111602-1G A111602-250MG A111602-25G A111602-50MG A111602-5G
77270-946EA 16.48 USD
77270-946 77270-948 77270-950 77270-952 77270-954
3,4',5-Trimethoxy-trans-stilbene 98%
3,4',5-Trimethoxy-trans-stilbene

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₃
MW: 270.33 g/mol
MDL Number: MFCD02093500
CAS Number: 22255-22-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR