6-Hydroxy-m-anisidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-methoxyphenol

A111599-100G A111599-10G A111599-1G A111599-250MG A111599-5G A111599-25G
77245-716EA 646.84 USD
77245-716 77245-718 77245-720 77245-722 77245-724 77549-166
6-Hydroxy-m-anisidine 97%
6-Hydroxy-m-anisidine

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MW: 139.16 g/mol
MDL Number: MFCD06616911
CAS Number: 20734-76-3
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR