(±)-Isonipecotic acid 98%

Supplier: Ambeed
Warning

A111595-100G A111595-10G A111595-25G A111595-500G A111595-1000G
77284-138EA 8 USD
77284-138 77284-140 77284-142 77284-144 77614-866
(±)-Isonipecotic acid 98%
(±)-Isonipecotic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MDL Number: MFCD00006004
CAS Number: 498-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR