1,3-Dibromo-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A111519-100G A111519-10G A111519-1G A111519-25G A111519-5G
77270-936EA 315.18 USD
77270-936 77270-938 77270-940 77270-942 77270-944
1,3-Dibromo-5-nitrobenzene 95%
1,3-Dibromo-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
MDL Number: MFCD00092529
CAS Number: 6311-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR