4'-Methyl-4-pentylbiphenyl 98%

Supplier: Ambeed
Warning

A111438-1G A111438-5G
77284-112EA 27.99 USD
77284-112 77284-114
4'-Methyl-4-pentylbiphenyl 98%
4'-Methyl-4-pentylbiphenyl

Precaución: for Research Use Only

Formula: C₁₈H₂₂
MDL Number: MFCD11053476
CAS Number: 64835-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR