6-(Pyridin-3-yl)pyrimidin-4-amine 97%

Supplier: Ambeed
Warning

A111369-100MG A111369-1G A111369-250MG
77257-682EA 39.14 USD
77257-682 77257-684 77257-686
6-(Pyridin-3-yl)pyrimidin-4-amine 97%
6-(Pyridin-3-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₉H₈N₄
MW: 172.19 g/mol
MDL Number: MFCD21234637
CAS Number: 1192814-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR