2-Bromo-4,5-dimethoxybenzonitrile 98%

Supplier: Ambeed
Warning

A111365-100MG A111365-250MG A111365-5G A111365-1G
77257-676EA 31.93 USD
77257-676 77257-678 77257-680 77745-824
2-Bromo-4,5-dimethoxybenzonitrile 98%
2-Bromo-4,5-dimethoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO₂
MW: 242.0693 g/mol
MDL Number: MFCD02256761
CAS Number: 109305-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR