PIK75 99%

Supplier: Ambeed
Warning

Synonyms: N'-((6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulphonohydrazide hydrochloride, PIK 75, (E)-N'-((6-bromoH-imidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulphonohydrazide hydrochloride, PIK-75 HCl, PIK75 Hydrochloride, PIK 75 Hydrochloride

A111266-10MG A111266-1MG A111266-25MG
77233-584EA 43.26 USD
77233-584 77233-586 77233-588
PIK75 99%
PIK75

Precaución: for Research Use Only

Formula: C₁₆H₁₅BrClN₅O₄S
MW: 488.74 g/mol
MDL Number: MFCD12546130
CAS Number: 372196-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR