Acotiamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Acotiamide HCl

A111223-100MG A111223-1MG
77245-712EA 35.02 USD
77245-712 77245-714
Acotiamide hydrochloride 98%
Acotiamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₃₁ClN₄O₅S
MW: 487.01 g/mol
MDL Number: MFCD23103502
CAS Number: 185104-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR