3-Iodo-4-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A111196-100MG A111196-1G A111196-250MG A111196-5G
77270-900EA 9.27 USD
77270-900 77270-902 77270-904 77270-906
3-Iodo-4-methoxybenzaldehyde 98%
3-Iodo-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.04 g/mol
MDL Number: MFCD01009500
CAS Number: 2314-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR