4,4'-Bipiperidine dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
A111182-1G
A111182-250MG
A111182-5G
77284-092EA
36.05
USD
77284-092
77284-094
77614-860
4,4'-Bipiperidine dihydrochloride 98%
4,4'-Bipiperidine dihydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂·2HCl MW: 241.21 g/mol |
MDL Number:
MFCD00005998 CAS Number: 78619-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...