4-Fluoro-(3-methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 4-Fluoro-3-methoxycarbonylphenylboronic acid

A111161-1G A111161-250MG A111161-25G A111161-5G
77233-576EA 23 USD
77233-576 77233-578 77233-580 77233-582
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid 98%
4-Fluoro-(3-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
MDL Number: MFCD06801688
CAS Number: 874219-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR