rel-Methyl (3R,4R)-1-benzyl-4-methylpyrrolidine-3-carboxylate 97%

Supplier: Ambeed
Warning

A111124-1G A111124-250MG A111124-5G A111124-10G
77257-668EA 19.57 USD
77257-668 77257-670 77257-672 77747-821
rel-Methyl (3R,4R)-1-benzyl-4-methylpyrrolidine-3-carboxylate 97%
rel-Methyl (3R,4R)-1-benzyl-4-methylpyrrolidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.31 g/mol
MDL Number: MFCD16660382
CAS Number: 473914-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR