6-Fluoro-m-anisaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-methoxybenzaldehyde, 6-Fluoro-3-anisaldehyde

A111001-100G A111001-10G A111001-1G A111001-25G A111001-5G
77233-560EA 305.01 USD
77233-560 77233-562 77233-564 77233-566 77233-568
6-Fluoro-m-anisaldehyde 98%
6-Fluoro-m-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD00070795
CAS Number: 105728-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR