3,5-Difluorophenylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (3,5-Difluorophenyl)hydrazine hydrochloride

A110877-100G A110877-10G A110877-1G A110877-25G A110877-5G
77284-058EA 191 USD
77284-058 77284-060 77284-062 77284-064 77284-066
3,5-Difluorophenylhydrazine hydrochloride 97%
3,5-Difluorophenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇Cl₃N₂
MDL Number: MFCD03094171
CAS Number: 502496-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR