2-Hydroxy-4-methoxyaniline hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxy-p-anisidine Hydrochloride, 2-Amino-5-methoxyphenol Hydrochloride
A110650-1G
A110650-250MG
A110650-25G
A110650-5G
77257-646EA
72.1
USD
77257-646
77257-648
77257-650
77257-652
2-Hydroxy-4-methoxyaniline hydrochloride 97%
2-Hydroxy-4-methoxyaniline hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₉NO₂ MW: 175.61 g/mol |
MDL Number:
MFCD00052002 CAS Number: 39547-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...